accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2NN

CAS RN 1622-57-7

IUPAC Name: 1-methyl-1H-1,3-benzodiazol-2-amine
Synonyms: 1-methyl-2-aminobenzimidazole
Molecular Weight (g/mol): 147.18
Molecular Formula: C8H9N3
InChi Key: XDFZKQJLNGNJAN-UHFFFAOYSA-N
SMILES: CN1C(N)=NC2=CC=CC=C12
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)