accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3H3CH3CCH3OHO

CAS RN 16225-26-6

IUPAC Name: 3,5-di-tert-butylbenzoic acid
Synonyms: 3,5-di-tert-butylbenzoic acid
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: NCTSLPBQVXUAHR-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(=C1)C(O)=O)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)