accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSOO

CAS RN 1623-93-4

IUPAC Name: [1,1'-biphenyl]-4-sulfonyl chloride
Synonyms: [1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol): 252.71
Molecular Formula: C12H9ClO2S
InChi Key: ALBQXDHCMLLQMB-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (4)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (5)
  • (4)
  • (3)