accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1624-01-7

IUPAC Name: dimethyl[tris(dimethylamino)silyl]amine
Synonyms: silanetetramine, octamethyl-
Molecular Weight (g/mol): 204.39
Molecular Formula: C8H24N4Si
InChi Key: SSCVMVQLICADPI-UHFFFAOYSA-N
SMILES: CN(C)[Si](N(C)C)(N(C)C)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results