accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 162401-62-9

IUPAC Name: 3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
Synonyms: 3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
Molecular Weight (g/mol): 258.22
Molecular Formula: C12H12F2O4
InChi Key: IGFDIFLMMLWKKY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(OCC2CC2)=C(OC(F)F)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results