accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16245-77-5

IUPAC Name: benzene-1,4-diamine; sulfuric acid
Synonyms: P-phenylenediamine sulfate
Molecular Weight (g/mol): 206.22
Molecular Formula: C6H10N2O4S
InChi Key: UFPKLWVNKAMAPE-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results