accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16260-48-3

IUPAC Name: 2-({3-[(2-hydroxyethyl)sulfanyl]propyl}sulfanyl)ethan-1-ol
Synonyms: 2-({3-[(2-hydroxyethyl)sulfanyl]propyl}sulfanyl)ethanol
Molecular Weight (g/mol): 196.32
Molecular Formula: C7H16O2S2
InChi Key: JIJWGPUMOGBVMQ-UHFFFAOYSA-N
SMILES: OCCSCCCSCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results