accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHSBr

CAS RN 162607-17-2

IUPAC Name: (5-bromothiophen-2-yl)boronic acid
Synonyms: 5-bromothiophen-2-ylboronic acid
Molecular Weight (g/mol): 206.85
Molecular Formula: C4H4BBrO2S
InChi Key: USJPOBDLWVCPGG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Br)S1
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results