accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 162607-18-3

IUPAC Name: (5-chlorothiophen-2-yl)boronic acid
Synonyms: 5-chlorothiophen-2-ylboronic acid
Molecular Weight (g/mol): 162.39
Molecular Formula: C4H4BClO2S
InChi Key: JFCLNCVCDFUJPO-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Cl)S1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)