accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHNNN

CAS RN 162640-98-4

IUPAC Name: 1-[4-(2H-1,2,3,4-tetrazol-5-yl)butyl]-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine
Synonyms: 1-[4-(2H-1,2,3,4-tetrazol-5-yl)butyl]-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine
Molecular Weight (g/mol): 397.53
Molecular Formula: C25H27N5
InChi Key: LQNGMDUIRLSESZ-UHFFFAOYSA-N
SMILES: C(CCC1=NNN=N1)CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results