accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHNOO

CAS RN 162648-54-6

IUPAC Name: 1-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)cyclohexane-1-carboxylic acid
Synonyms: 1-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 365.43
Molecular Formula: C22H23NO4
InChi Key: VCIVAWBKUQJNSX-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)