accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3COFFF

CAS RN 1628442-55-6

IUPAC Name: 2-[4-(difluoromethoxy)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[4-(difluoromethoxy)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 288.07
Molecular Formula: C13H16BF3O3
InChi Key: LLJRSXMGPKHUQX-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C=C(OC(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results