accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONNN

CAS RN 162848-16-0

IUPAC Name: 4-(1H-1,2,4-triazol-1-yl)benzoate
Synonyms: 4-(1,2,4-triazol-1-yl)benzoate
Molecular Weight (g/mol): 188.17
Molecular Formula: C9H6N3O2
InChi Key: FOMQQGKCPYKKHQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1)N1C=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results