accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16298-03-6

IUPAC Name: methyl 3-aminopyrazine-2-carboxylate
Synonyms: methyl 3-aminopyrazine-2-carboxylate
Molecular Weight (g/mol): 153.14
Molecular Formula: C6H7N3O2
InChi Key: INCSQLZZXBPATR-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC=CN=C1N
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)