accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16303-58-5

IUPAC Name: 5,5'-dimethyl-2,2'-bithiophene
Synonyms: 5,5'-dimethyl-2,2'-bithiophene
Molecular Weight (g/mol): 194.31
Molecular Formula: C10H10S2
InChi Key: DKHDQTBBXQMFPP-UHFFFAOYSA-N
SMILES: CC1=CC=C(S1)C1=CC=C(C)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results