accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NHO

CAS RN 163061-74-3

IUPAC Name: 1-amino-2,3-dihydro-1H-inden-2-ol
Synonyms: 1-amino-2,3-dihydro-1H-inden-2-ol
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: LOPKSXMQWBYUOI-UHFFFAOYNA-N
SMILES: NC1C(O)CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)