accessibility menu, dialog, popup

UserName

Viewing profile as user:
OON

CAS RN 1631-25-0

IUPAC Name: 1-cyclohexyl-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-cyclohexylpyrrole-2,5-dione
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: BQTPKSBXMONSJI-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1CCCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)