accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1631-83-0

IUPAC Name: chlorodiphenylsilyl
Synonyms: chlorodiphenylsilyl
Molecular Weight (g/mol): 217.75
Molecular Formula: C12H10ClSi
InChi Key: YCITZMJNBYYMJO-UHFFFAOYSA-N
SMILES: Cl[Si](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)