accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1632-16-2

IUPAC Name: 3-methylideneheptane
Synonyms: 2-ethyl-1-hexene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: XTVRLCUJHGUXCP-UHFFFAOYSA-N
SMILES: CCCCC(=C)CC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results