accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1632-73-1

IUPAC Name: (1S,2S,4R)-1,3,3-trimethylbicyclo[2.2.1]heptan-2-ol
Synonyms: fenchyl alcohol (1r-endo)-fenchol (1s-endo)-fenchol α-fenchol ((endo)-(+-))-fenchol (endo)-isomer fenchol fenchol, (exo)- fenchol, (1R-endo)-isomer fenchol, ((endo)-(+-))- fenchol, (endo)-
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: IAIHUHQCLTYTSF-MRTMQBJTSA-N
SMILES: CC1(C)[C@@H]2CC[C@@](C)(C2)[C@@H]1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results