accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1632075-13-8

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-4-[(triphenylmethyl)carbamoyl]butanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}-4-(triphenylmethylcarbamoyl)butanoic acid
Molecular Weight (g/mol): 624.74
Molecular Formula: C40H36N2O5
InChi Key: RMAUSWDYZJQCPJ-BHVANESWSA-N
SMILES: CN([C@@H](CCC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)