accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONOOH3C

CAS RN 163229-48-9

IUPAC Name: 1-tert-butyl 2-methyl 1H-indole-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl indole-1,2-dicarboxylate
Molecular Weight (g/mol): 275.3
Molecular Formula: C15H17NO4
InChi Key: UJVYYFYNECMKRA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=CC=CC=C2N1C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results