accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 163252-36-6

IUPAC Name: 1-[(2S,3R,4S,5S)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: clevudine
Molecular Weight (g/mol): 260.22
Molecular Formula: C10H13FN2O5
InChi Key: GBBJCSTXCAQSSJ-XQXXSGGOSA-N
SMILES: CC1=CN([C@H]2O[C@@H](CO)[C@H](O)[C@H]2F)C(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results