accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOClFFF

CAS RN 163295-75-8

IUPAC Name: [4-(trifluoromethyl)phenyl]methanesulfonyl chloride
Synonyms: [4-(trifluoromethyl)phenyl]methanesulfonyl chloride
Molecular Weight (g/mol): 258.64
Molecular Formula: C8H6ClF3O2S
InChi Key: KKBNUPMMAGEQAT-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(CS(Cl)(=O)=O)C=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results