accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONNOHOHOOOHO

CAS RN 1633-00-7

IUPAC Name: 2-({6-[bis(carboxymethyl)amino]hexyl}(carboxymethyl)amino)acetic acid
Synonyms: ({6-[bis(carboxymethyl)amino]hexyl}(carboxymethyl)amino)acetic acid
Molecular Weight (g/mol): 348.35
Molecular Formula: C14H24N2O8
InChi Key: YGDVXSDNEFDTGV-UHFFFAOYSA-N
SMILES: OC(=O)CN(CCCCCCN(CC(O)=O)CC(O)=O)CC(O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results