accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1633-78-9

IUPAC Name: 6-sulfanylhexan-1-ol
Synonyms: 6-sulfanylhexan-1-ol
Molecular Weight (g/mol): 134.24
Molecular Formula: C6H14OS
InChi Key: UGZAJZLUKVKCBM-UHFFFAOYSA-N
SMILES: OCCCCCCS
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)