accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2(S)H2COHO

CAS RN 16338-48-0

IUPAC Name: (2S)-2-aminopent-4-enoic acid
Synonyms: (2S)-2-aminopent-4-enoic acid
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: WNNNWFKQCKFSDK-BYPYZUCNSA-N
SMILES: N[C@@H](CC=C)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)