accessibility menu, dialog, popup

UserName

Viewing profile as user:
SiSi

CAS RN 16343-18-3

IUPAC Name: tetraphenyldisilene
Synonyms: tetraphenyldisilene
Molecular Weight (g/mol): 364.59
Molecular Formula: C24H20Si2
InChi Key: BXLNWOAYQXBHCY-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Si](C1=CC=CC=C1)=[Si](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results