accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOO

CAS RN 1635-31-0

IUPAC Name: 3-amino-2H-chromen-2-one
Synonyms: 3-aminochromen-2-one
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: QWZHDKGQKYEBKK-UHFFFAOYSA-N
SMILES: NC1=CC2=CC=CC=C2OC1=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results