CAS RN 1635-84-3
CAS RN 1635-84-3
IUPAC Name:
2,4-dimethyl-6-nitroaniline
Synonyms:
2,4-dimethyl-6-nitroaniline
Molecular Weight (g/mol):
166.18
Molecular Formula:
C8H10N2O2
InChi Key:
VSRYYONYIUUFFY-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(N)C(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals 4,6-Dimethyl-2-nitroaniline, 97%
CAS: 1635-84-3 Molecular Formula: C8H10N2O2 Molecular Weight (g/mol): 166.18 InChI Key: VSRYYONYIUUFFY-UHFFFAOYSA-N Synonym: 4,6-dimethyl-2-nitroaniline,6-nitro-2,4-xylidine,2,4-dimethyl-6-nitrophenyl amine,benzenamine, 2,4-dimethyl-6-nitro,2,4-dimethyl-6-nitrophenylamine,acmc-209uyq,intermediates-zcf02230,4-amino-5-nitro-m-xylene,cambridge id 5100320,2-nitro-4,6-dimethylaniline PubChem CID: 74219 IUPAC Name: 2,4-dimethyl-6-nitroaniline SMILES: CC1=CC(=C(C(=C1)[N+](=O)[O-])N)C