accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 163520-33-0

IUPAC Name: ethyl 5,5-diphenyl-4,5-dihydro-1,2-oxazole-3-carboxylate
Synonyms: ethyl 5,5-diphenyl-4H-1,2-oxazole-3-carboxylate
Molecular Weight (g/mol): 295.34
Molecular Formula: C18H17NO3
InChi Key: MWKVXOJATACCCH-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NOC(C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results