accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNN

CAS RN 16357-69-0

IUPAC Name: 4-aminopyrimidine-5-carbonitrile
Synonyms: 4-aminopyrimidine-5-carbonitrile
Molecular Weight (g/mol): 120.12
Molecular Formula: C5H4N4
InChi Key: JAIYUIOGVNRXEW-UHFFFAOYSA-N
SMILES: NC1=NC=NC=C1C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results