accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 163596-75-6

IUPAC Name: 4-bromo-3-nitrobenzaldehyde
Synonyms: 4-bromo-3-nitrobenzaldehyde
Molecular Weight (g/mol): 230.02
Molecular Formula: C7H4BrNO3
InChi Key: SAFSVELFSYQXOV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Br)C=CC(C=O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results