accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHOOHO

CAS RN 1636-27-7

IUPAC Name: dipropylpropanedioic acid
Synonyms: dipropylpropanedioic acid
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: DIRSQLKNZQKDBK-UHFFFAOYSA-N
SMILES: CCCC(CCC)(C(O)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results