accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 163680-77-1

IUPAC Name: 6-(1-aminocyclopropyl)-7-fluoro-2-methyl-10-oxo-4-oxa-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid; methanesulfonic acid
Synonyms: methanesulfonic acid; pazufloxacin
Molecular Weight (g/mol): 414.4
Molecular Formula: C17H19FN2O7S
InChi Key: UDHGFPATQWQARM-UHFFFAOYNA-N
SMILES: CS(O)(=O)=O.CC1COC2=C3N1C=C(C(O)=O)C(=O)C3=CC(F)=C2C1(N)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results