accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNCH3

CAS RN 16382-15-3

IUPAC Name: ethyl 5-methyl-1H-indole-2-carboxylate
Synonyms: ethyl 5-methyl-1H-indole-2-carboxylate
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: KMVFKXFOPNKHEM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(C)=CC=C2N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results