accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16386-93-9

IUPAC Name: 2,2-dimethylpent-4-enoic acid
Synonyms: 2,2-dimethylpent-4-enoic acid
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: BGUAPYRHJPWVEM-UHFFFAOYSA-N
SMILES: CC(C)(CC=C)C(O)=O
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)