accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1638847-77-4

IUPAC Name: methyl 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Synonyms: methyl 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular Weight (g/mol): 280.1
Molecular Formula: C14H18BFO4
InChi Key: LKZMOGYZLFRTQN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(B2OC(C)(C)C(C)(C)O2)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results