accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1639-09-4

IUPAC Name: heptane-1-thiol
Synonyms: 1-heptanethiol
Molecular Weight (g/mol): 132.27
Molecular Formula: C7H16S
InChi Key: VPIAKHNXCOTPAY-UHFFFAOYSA-N
SMILES: CCCCCCCS
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)