accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3C

CAS RN 1640-39-7

IUPAC Name: 2,3,3-trimethyl-3H-indole
Synonyms: 2,3,3-trimethylindole
Molecular Weight (g/mol): 159.23
Molecular Formula: C11H13N
InChi Key: FLHJIAFUWHPJRT-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=CC=C2C1(C)C
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)