accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 164033-12-9

IUPAC Name: (1-isocyanatopropyl)benzene
Synonyms: (1-isocyanatopropyl)benzene
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: XGPXXSJFZSZULR-UHFFFAOYNA-N
SMILES: CCC(N=C=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results