accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSNOOH

CAS RN 16405-98-4

IUPAC Name: 2,1,3-benzothiadiazole-5-carboxylic acid
Synonyms: 2,1,3-benzothiadiazole-5-carboxylic acid
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H4N2O2S
InChi Key: YHMXJZVGBCACMT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=NSN=C2C=C1
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results