accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16409-43-1

IUPAC Name: 4-methyl-2-(2-methylprop-1-en-1-yl)oxane
Synonyms: rose oxide laevo-rose oxide isobutenyl methyltetrahydropyran
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: CZCBTSFUTPZVKJ-UHFFFAOYNA-N
SMILES: CC1CCOC(C1)C=C(C)C
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)