accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1642-81-5

IUPAC Name: 4-(chloromethyl)benzoic acid
Synonyms: 4-(chloromethyl)benzoic acid
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: OITNBJHJJGMFBN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CCl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)