accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHCH3

CAS RN 16423-19-1

IUPAC Name: 4,8a-dimethyl-decahydronaphthalen-4a-ol
Synonyms: octahydro-dimethyl-4α-naphthol
Molecular Weight (g/mol): 182.31
Molecular Formula: C12H22O
InChi Key: JLPUXFOGCDVKGO-UHFFFAOYNA-N
SMILES: CC1CCCC2(C)CCCCC12O
Concentration 
  • (2)
Grade 
  • (2)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)