accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16423-54-4

IUPAC Name: 5-chloro-2-(methylsulfanyl)aniline
Synonyms: 5-chloro-2-(methylsulfanyl)aniline
Molecular Weight (g/mol): 173.66
Molecular Formula: C7H8ClNS
InChi Key: RWUQNIPYCAHKMO-UHFFFAOYSA-N
SMILES: CSC1=C(N)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results