accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOBrNOO

CAS RN 16426-64-5

IUPAC Name: 2-bromo-4-nitrobenzoic acid
Synonyms: 2-bromo-4-nitrobenzoic acid
Molecular Weight (g/mol): 246.02
Molecular Formula: C7H4BrNO4
InChi Key: CEXGTXNIIFSPSF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1Br)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results