accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1643-20-5

IUPAC Name: N,N-dimethyldodecanamine oxide
Synonyms: lauryl dimethylamine-N-oxide
Molecular Weight (g/mol): 229.41
Molecular Formula: C14H31NO
InChi Key: SYELZBGXAIXKHU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC[N+](C)(C)[O-]
Form 
  • (2)
Product Type 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results