CAS RN 1643-26-1
IUPAC Name:
3-(2-fluorophenyl)propanoate
Synonyms:
3-(2-fluorophenyl)propanoate
Molecular Weight (g/mol):
167.16
Molecular Formula:
C9H8FO2
InChi Key:
GUZLQEOSDXLCKX-UHFFFAOYSA-M
SMILES:
[O-]C(=O)CCC1=CC=CC=C1F
1
–
1
of
1
results
1
Promotions Available
3-(2-Fluorophenyl)propionic acid, 96%
3-(2-Fluorophenyl)propionic acid, 96%
CAS: 1643-26-1 Molecular Formula: C9H9FO2 Molecular Weight (g/mol): 168.167 MDL Number: MFCD01310820 InChI Key: GUZLQEOSDXLCKX-UHFFFAOYSA-N PubChem CID: 2063866 IUPAC Name: 3-(2-fluorophenyl)propanoic acid SMILES: C1=CC=C(C(=C1)CCC(=O)O)F