accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1643-30-7

IUPAC Name: 3-(4-bromophenyl)propanoic acid
Synonyms: 3-(4-bromophenyl)propanoic acid
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: NCSTWHYWOVZDOC-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results